Lab Seal™ Tamper-Evident Specimen Bag, Elkay Plastics
Catalog #: 10789-538
Supplier: Elkay Plastics
Specifications
- Dimensions:10×10"
- Color:Clear
- Description:3-Wall unprinted bag without removable biohazard symbol
- Thickness:1.8 mil
- Cat. no.:10789-538
- Supplier No.:LABA1010
Specifications
About this item
Elkay’s Lab-Seal™ Tamper-Evident specimen bags with Seal-N-Rip™ removable biohazard symbol provide an means for securely transferring specimens in hospital and medical environments while helping to save on disposal costs.
- A special mechanism where once its marked tab is pulled, the bag will seal securely and permanently with adhesive glue
Once the bag is sealed, the only way to access the contents would be to tear off the top indicated by a tear line, which would reveal any tampering.
The bags have a special removable biohazard symbol which can be torn off and allows the bags to be disposed of with regular waste if they are uncontaminated.
Customers Who Bought This Also Bought