To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Animal-free:Y
- Protease-free:Y
- Protein Function:Cytokine
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Mouse
- Size:5 μg
- Storage conditions:Store at –20 °C prior to opening
- Endotoxin content:≤ 1.00 EUs/μg
- Biological activity:Induce IL-6 production from M1 cells at less than or equal to 1 ng/mL or (1 x 10^6 units/mg specific units)
- Reconstitution instructions:Reconstitute with Sterile 10 mM acetic acid at 0.1 mg/mL
- Protein synonyms:Leukocyte Inhibitory Factor
- UniProtKB:P09056
- Protein/peptide name:LIF
- Purity:>95%
- Molecular weight:20.0 (kDa) Monomer
- Sequence:MSPLPITPVN ATCAIRHPCH GNLMNQIKNQ LAQLNGSANA LFISYYTAQG EPFPNNVEKL CAPNMTDFPS FHGNGTEKTK LVELYRMVAY LSASLTNITR DQKVLNPTAV SLQVKLNATI DVMRGLLSNV LCRLCNKYRV GHVDVPPVPD HSDKEAFQRK KLGCQLLGTY KQVISVVVQA F
- Endotoxin level:Low
- Formulation:Lyophilized from a sterile (0.2 micron) filtered aqueous solution containing 0.1% Trifluoroacetic Acid (TFA)
- Shipping temperature:Ambient
- Cat. no.:10788-080
- Origin:USA
- Supplier No.:200-33AF-5UG
Specifications
About this item
Mouse leukemia inhibitory factor (LIF) is a member of the interleukin 6 (IL-6) family that is made by a variety of adult and embryonic tissues. LIF signals through the glycoprotein 130 (gp130)/LIF receptor (LIFR) heterodimer to activate STAT3 and MAPK signaling. LIF functions during hematopoietic differentiation, neuronal cell differentiation, kidney development, and inflammatory processes. The application of mouse LIF to long-term culture systems promotes mouse embryonic stem (ES) cell self-renewal and pluripotency, similar to the functional activity of bFGF (FGF2) in human ES cell culture systems.
- Animal-free
Shenandoah Biotechnology is a manufacturer of research grade recombinant proteins typically used as cytokines, growth factors and chemokines in cell based assays and cell culture. Each production lot is tested for QA and the expected values for each product can be viewed under the product specifications tab.
A COA is supplied with each product shipped detailing purity, formulation, biological activity, endotoxin levels, reconstitution instructions and recommended storage conditions. This product is for research use only and is not for use in humans.