Order Entry
ContactUsLinkComponent
Recombinant Human IL-4
Human Recombinant IL-4 (from E. coli)
Catalog #: 10779-606
Supplier:  Peprotech
CAS Number:  
Human Recombinant IL-4 (from E. coli)
Catalog #: 10779-606
Supplier:  Peprotech
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Gene ID:
    3565 P05112
  • Protease-free:
    Y
  • Protein Function:
    Cytokines and Growth Factors
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Conjugation:
    Unconjugated
  • Size:
    20 μg
  • Storage conditions:
    Store at –20 °C prior to reconstitution.
  • Endotoxin content:
    >0.1 ng/μg
  • Biological activity:
    Biological Activity tests can vary. Please refer to the Product CoA for information.
  • Form:
    Lyophilized
  • Reconstitution instructions:
    Reconstitution instructions can vary. Please refer to the product CoA for info.
  • Protein synonyms:
    B-cell Stimulating Factor (BSF-1)|BCGF|BCDF|Interleukin-4
  • Protein/peptide name:
    IL-4
  • Purity:
    0.98
  • Molecular weight:
    15.1 kDa
  • Sequence:
    MHKCDITLQE IIKTLNSLTE QKTLCTELTV TDIFAASKNT TEKETFCRAA TVLRQFYSHH EKDTRCLGAT AQQFHRHKQL IRFLKRLDRN LWGLAGLNSC PVKEANQSTL ENFLERLKTI MREKYSKCSS
  • Endotoxin level:
    Low
  • Formulation:
    Formulation can vary. Please refer to product CoA for information.
  • Shipping temperature:
    Ambient
  • Cat. no.:
    10779-606
  • Origin:
    USA
  • Supplier No.:
    200-04-20UG

Specifications

About this item

IL-4 is a pleiotropic cytokine that regulates diverse T and B cell responses including cell proliferation, survival and gene expression. Produced by mast cells, T cells and bone marrow stromal cells, IL-4 regulates the differentiation of naive CD4+ T cells into helper Th2 cells, characterized by their cytokine-secretion profile that includes secretion of IL-4, IL-5, IL-6, IL-10, and IL-13, which favor a humoral immune response. Another dominant function of IL-4 is the regulation of immunoglobulin class switching to the IgG1 and IgE isotypes. Excessive IL-4 production by Th2 cells has been associated with elevated IgE production and allergy. Recombinant Human IL-4 is a 15.1 kDa globular protein containing 130 amino acid residues.

Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.
Customers Who Bought This Also Bought