Order Entry
ContactUsLinkComponent
Recombinant Human IL-8 (72 a
Human Recombinant IL-8 (from E. coli)
Catalog #: 10772-958
Supplier:  Peprotech
CAS Number:  
Human Recombinant IL-8 (from E. coli)
Catalog #: 10772-958
Supplier:  Peprotech
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Gene ID:
    3576 P10145
  • Protease-free:
    Y
  • Protein Function:
    Cytokines and Growth Factors
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Conjugation:
    Unconjugated
  • Size:
    25 μg
  • Storage conditions:
    Store at –20 °C prior to reconstitution.
  • Endotoxin content:
    >0.1 ng/μg
  • Biological activity:
    Biological Activity tests can vary. Please refer to the Product CoA for information.
  • Form:
    Lyophilized
  • Reconstitution instructions:
    Reconstitution instructions can vary. Please refer to the product CoA for info.
  • Protein synonyms:
    Neutrophil Activating Factor (NAF)|Interleukin-8|NAP-1|Monocyte-Derived Neutrophil Chemotactic Factor (MDNCF)
  • Protein/peptide name:
    IL-8
  • Purity:
    0.98
  • Molecular weight:
    8.4 kDa
  • Sequence:
    SAKELRCQCI KTYSKPFHPK FIKELRVIES GPHCANTEII VKLSDGRELC LDPKENWVQR VVEKFLKRAE NS
  • Endotoxin level:
    Low
  • Formulation:
    Formulation can vary. Please refer to product CoA for information.
  • Shipping temperature:
    Ambient
  • Cat. no.:
    10772-958
  • Origin:
    USA
  • Supplier No.:
    200-08M-25UG

Specifications

About this item

IL-8 is a proinflammatory CXC chemokine that can signal through the CXCR1 and CXCR2 receptors. It is secreted by monocytes and endothelial cells. IL-8 chemoattracts and activates neutrophils. Recombinant Human IL-8 (CXCL8) (monocyte-derived) is an 8.4 kDa protein containing 72 amino acid residues.

Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.
Customers Who Bought This Also Bought