VWR® Round Carboy, Narrow Mouth, Sterile
Catalog #: 10755-121
Supplier: VWR International
Specifications
- Capacity:20 L
- Description:Carboy, Round, Sterile, Narrow Mouth
- Material:HDPE
- Neck I.D.:83 mm
Specifications
About this item
Use for storing and dispensing solutions and media. Ideal for sterile water storage. These high-density polyethylene, disposable carboys are ideal for storing and transfer of sterile fluids and pharmaceutical/biotech reagents.
- Sterile
- HDPE formats available
- Excellent chemical resistance
- Clear graduation marks
- Narrow-mouth
- Feature an integrated shoulder handle
Carboys are well suited for use in temperatures ranging from -100 to 100 °C (-148 to 212 °F). Carboys are leak-proof under standard test conditions.
HDPE carboys are not autoclavable.
Certifications: Resin meets US FDA 21 CFR, USP Class VI.
Customers Who Bought This Also Bought