Order Entry
ContactUsLinkComponent
Weigh boat
VWR® Disposable Hexagonal Weigh Boats
Catalog #: 10770-458
Supplier:  VWR International
Weigh boat
VWR® Disposable Hexagonal Weigh Boats
Catalog #: 10770-458
Supplier:  VWR International

Specifications

  • Size:
    X-Large
  • Top I.D.:
    127 mm (5")
  • Base I.D.:
    89 mm (3½")

Specifications

About this item

These polystyrene weighing boats feature smooth surfaces to enable accurate pour-out with minimal sample loss.

  • Flat bottom avoids tipping
  • Sloping sides provide accurate transfer
  • Antistatic
  • Temperature resistant up to 70°C (158°F)

Biologically inert and resistant to dilute and weak acids, aqueous solutions, alcohols and bases. These flexible, antistatic boats bend at points to form pouring spouts.

Customers Who Bought This Also Bought