ContactUsLinkComponent
Rack
SP Bel-Art Magnetic Bead Separation Racks, Bel-Art Products, a part of SP
Catalog #: 10770-238
Rack
SP Bel-Art Magnetic Bead Separation Racks, Bel-Art Products, a part of SP
Catalog #: 10770-238
Specifications
  • Cat. no.:
    10770-238
  • Fits:
    10×1.5 - 2.0 ml microcentrifuge tubes
  • Supplier No.:
    F19900-0002
Specifications
About this item
Designed for use in all magnetic separation experiments, these powerful racks sequester magnetic beads so that affinity ligands (antibodies, streptavidin, or proprietary reagents for coupling nucleic acids) or rinse solutions can be added or decanted in assays.
  • Powerful magnets quickly secure beads
  • Spring plunger on tube sleeves
  • Simply turn tube for efficient bead mixing
  • Racks fit microplates following ANSI standards
  • Clear acrylic allows unobstructed view of tube contents
Magnetic Bead Separation Racks offer exceptional quality and value compared to competing brands
96-Well PCR plate model has 24 magnets that snugly hold the plate while attracting beads to the wall of the tubes.
Flat bottom microplate model has magnets covering entire footprint to accept all standard micro-plates with any number of wells; 6, 12, 24, 48, 96, etc.
Patent pending, independent sleeves on 50 mL and 15/5 mL models make liquid addition and removal easier with less accidental fluid loss.
Customers Who Bought This Also Bought