Fume Extractor
Catalog #: CA10770-070
Supplier: Movex
Customers Who Bought This Also Bought
Barium chloride dihydrate ≥99.0% ACS, VWR Chemicals BDH®
Supplier: VWR International
184,95 $ to 1 807,94 $
Puritan® Tapered Mini Cotton Tipped Applicator, Wood Handle, Puritan Medical Products
Supplier: Puritan Medical Products
62,97 $ to 529,70 $
Potassium nitrate ≥99.0%, crystallised ACS, VWR Chemicals BDH®
Supplier: VWR International
179,52 $ to 927,60 $
Alcohol ≥70%, Reagent Grade (denatured with methanol and isopropanol), VWR Chemicals BDH®
Supplier: VWR International
131,08 $ to 405,43 $