Potassium nitrate (nitre- KNO₃)
Catalog #: 80705-020
Supplier: Avantor
CAS Number: 7757-79-1
Specifications
- Packaging:Bag
- Size:25 kg
- Cat. no.:80705-020
- Supplier No.:AMAS029-942-01
Specifications
About this item
Synonyms: Nitrate of Potash
Melting Pt:334 °C
Density:2.109 g/cm³ (20 °C)
ADR:5.1,III
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Melting Pt: 334 °C
:Flash Pt:
:Flash Pt:
:M.W. 101.1 g/mol
Merck Index:12,07815
:Potassium nitrate
Storage Temperature:Ambient
EINECS:231-818-8
UN:1486
REACH:01-2119488224-35
:5.1
:III
:GHS03
Customers Who Bought This Also Bought