Silica flour (quartz), flour 200 mesh
Catalog #: 80705-014
Supplier: Avantor
CAS Number: 112945-52-5
Specifications
- Packaging:Bag
- Size:22.7 kg
- Cat. no.:80705-014
- Supplier No.:AMASMR115-22.7KG
Specifications
About this item
:Silica flour (quartz)
Boiling Pt:2230 °C (1013 hPa)
Melting Pt:1427 °C
Density:2.622 - 2.645 g/cm³ (20 °C)
:Length:
:Width:
:Height:
:Ø
:Thickness:
:200 mesh
:Boiling Pt:
:Boiling Pt: 2230 °C (1013 hPa)
:Melting Pt:
:Melting Pt: 1427 °C
:Flash Pt:
:Flash Pt:
:M.W. 60.083 g/mol
Storage Temperature:Ambient
EINECS:238-878-4
:GHS08
Customers Who Bought This Also Bought