Spectra/Por® 1-5 Standard RC Dry Dialysis Trial Kits, Spectrum® Laboratories
Catalog #: 10768-530
Supplier: Spectrum Laboratories
Specifications
- Ø:14.6 mm
- Length:5 m
- Volume/Length:1.7 ml/cm
- Cat. no.:10768-530
- MWCO:6 - 8 kD
- Flat Width:23 mm
- Supplier No.:132650T
Specifications
About this item
Ideal for small volume dialysis or evaluation of membrane for an application, multi-purpose standard Regenerated Cellulose (RC) tubing is now available in 1 and 5 m length trial kits with a pair of tubing closures.
- Hydrophilic, symmetric porosity
Standard Regenerated Cellulose (RC) is composed of natural cellulose regenerated from cotton linters, these membranes are perfectly suited for a broad range of laboratory dialysis. This clear and flexible membrane type with a symmetric porosity is economical and sturdy, ideal for desalting, buffer exchanges, macromolecular purifications and other general applications.
Delivery: Kit includes 5 m of Spectra/Por 1, 2, 3, 4, or 5 dialysis tubing and 2 closures.
Customers Who Bought This Also Bought