Filter Discs, Screen Support, Ace Glass Incorporated
Catalog #: 10767-248
Supplier: Ace Glass
Small Business Enterprise
Specifications
- Description:Fabric Screen Material (by the Yard), 295 micron
- Cat. no.:10767-248
- Supplier No.:5814-107
Specifications
About this item
These filter discs have a wide variety of uses.
Available in highly absorbent paper, polypropylene, nylon, or PTFE.
Ordering information: For filter discs made from polyethylene, fluorocarbon, or other materials, please contact your Avantor representative.
Customers Who Bought This Also Bought