ReadyRack™ Tryptic Soy Broth (TSB), Disposable Tube Rack
Catalog #: CA10436-342
Supplier: HARDY DIAGNOSTICS CA
CAS Number:
Specifications
- Packaging:80 per package
- Description:TSB, USP in ReadyRack™
- Size:16×125 mm
- Fill Volume:10 ml
- Application:Breweries, Wineries and distilleries, Cosmetics, Pharmaceutical manufacturing
- Supplier No.:K82BX
Specifications
About this item
Tryptic Soy Broth (TSB) in a ReadyRack™ is for sterility testing and cultivation of microorganisms. TSB is also known as Soybean-Casein Digest Broth, USP.
- For sterility testing in cleanrooms
- Can be rinsed with alcohol
- Avoid contamination
The ReadyRack is a plastic rack designed for cleanroom use and can be rinsed with alcohol.
Simply remove ReadyRack from box outside of cleanroom. Spray down the plastic bag with isopropyl alcohol, and allow to dry. Take into cleanroom and remove plastic bag. Spray down entire ReadyRack with isopropyl alcohol, and allow to dry.
It is a convenient way to bring media into a controlled area while avoiding outside contamination.
Customers Who Bought This Also Bought