Order Entry
ContactUsLinkComponent
Foil tape
Hareta Replacement Slide Hot Foil Tape, Springside Scientific
Catalog #: 10756-188
Foil tape
Hareta Replacement Slide Hot Foil Tape, Springside Scientific
Catalog #: 10756-188
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Dimensions:
    25 mm x 300 m
  • Cat. no.:
    10756-188
  • Packaging:
    Round Metal Box
  • Supplier No.:
    HSFT300

Specifications

About this item

This tape is used for automated slide printing and designed for use on Thermo SlideMate printers to replace the expensive tape currently offered by the OEM.

Prints up to 12,000 slides per roll. Will not fade, even in the harshest processing environments, including alcohols and xylene. A direct replacement for the original OEM tape.

Packaging: Each roll is 25 mm x 300 m in length and is a long lasting, standard sized 984' roll.
Customers Who Bought This Also Bought