Order Entry
ContactUsLinkComponent
Paraffin repellent liquid
Paraffin Repellent Kit, Springside Scientific
Catalog #: 10756-182
Paraffin repellent liquid
Paraffin Repellent Kit, Springside Scientific
Catalog #: 10756-182
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    Paraffin Repellent Liquid Bottle
  • Size:
    4 oz.
  • Cat. no.:
    10756-182
  • Supplier No.:
    PRCM01

Specifications

About this item

The Paraffin Repellent Kit works to prevent wax from sticking to painted surfaces, can be used as an effective mold release, and will clean up wax from instruments and counter tops.

  • All natural and non-toxic
  • Cucumber melon scented
Ordering information: Each kit includes four bottles (4oz. each) and one plastic scraper. Individual bottles are available separately.
Customers Who Bought This Also Bought