To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Description:Pocket Pro+, pH
- Operating temperature range:0…50 °C
- Battery life:>280 hrs
- Sensor type:Single-Junction (electrode/ sensor)
- pH accuracy:±0.02 pH
- pH calibration:3 points; customer-defined 2 point
- pH range:0.0 - 14.0
- pH resolution:0.1
- Battery type:AAA×4
- Cat. no.:CA89508-374
- Supplier No.:9532000
Specifications
About this item
Hach Pocket Pro and Pro+ are engineered to deliver accurate results. Calibration functions for ongoing testing, diagnostics with select models and a stabilized sample lock ensure you get the right reading. You also get replaceable batteries for convenient field use, and a large, easy-to-read LCD screen.
Confidence in results; to maximize accuracy all testers are factory calibrated, select models offer user-calibration, and pH models have performance diagnostics.
Auto-Buffer Recognition. ATC. Selectable Temp (°C or °F). Auto-Off. Reading Hold. Waterproof (IP 67 and floats). Quick-Release Lanyard (included). Transparent Sensor Cap.
Save time and money; easy-to-replace AAA batteries and convenient sample cup save time on maintenance and measurements. Ease of use; intuitive user interface guides users through the calibration and measurement process with a large, easy-to-read LCD screen.