Order Entry
ContactUsLinkComponent
Electrode
IntelliCAL™ MTC101 Standard Gel Filled ORP Electrodes, Hach
Catalog #: CA89501-162
Supplier:  Hach
Electrode
IntelliCAL™ MTC101 Standard Gel Filled ORP Electrodes, Hach
Catalog #: CA89501-162
Supplier:  Hach
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    IntelliCAL™ MTC101 Standard ORP Electrode, 3 m cable
  • Material:
    Epoxy
  • Cat. no.:
    CA89501-162
  • Supplier No.:
    MTC10103

Specifications

About this item

IntelliCAL™ MTC101 is a digital, combination oxidation reduction potential (ORP/Redox) probe with a non-refillable, gel-electrolyte reference and built-in temperature sensor.

-Provides fast, stable response in a variety of applications
-Ultimate traceability in measurement history
-User alerted when re-calibration is needed
-Can be moved between meters without the need to re-calibrate or re-enter measurement settings

  • Provides fast, stable response in a variety of applications
  • Ultimate traceability in measurement history
  • User alerted when recalibration is needed
  • Flat disc sensor for easy cleaning
  • Low maintenance
  • Ag/AgCl reference system
  • Can be moved between meters without the need to recalibrate or reenter measurement settings

IntelliCAL™ MTC101 is a digital, combination oxidation reduction potential (ORP/Redox) probe with a non-refillable, gel-electrolyte reference and built-in temperature sensor.

The standard version is available with a 1 or 3 meter cable and is intended for laboratory use. The MTC101 is ideal for measuring absolute mV values in wastewater, drinking water and general applications.

Weight: 0.17 lbs. (0.076 kg)

Customers Who Bought This Also Bought