Specifications
- Description:Ultrasnap™ Surface ATP Swabs (box of 100)
- Cat. no.:CA10142-318
- Supplier No.:US2020
Specifications
About this item
SystemSURE Plus™ is the next-generation system of the SystemSURE II ATP hygiene monitoring system. With improved functionality, new software, and increased memory, the SystemSURE Plus meets the demands of the largest and smallest companies around the world.
- Provides real-time feedback and facilitates long-term data analysis
- Optimizes and verifies that cleaning procedures are working
- Assists in developing and improving process and risk assessment programs (ie. HACCP & GMPs)
- No scheduled yearly maintenance is necessary
- Powered by two AA batteries
The SystemSURE Plus™ is designed for convenient, single-hand use to make using the system and navigating the menus easy for every user. Don't let the petite shape fool you - SystemSURE Plus™ makes no compromises when it comes to durability. The rugged outer shell and splash-proof keypad keep SystemSURE Plus safe in the harshest environments. For extra protection, every system comes with a complimentary carrying case with a shoulder strap for hands-free transportation to test locations in your facility.
A clean production environment is a key component to producing safe, quality foods. By incorporating ATP sanitation monitoring into preoperative equipment inspection and change-over cleaning verification, quality personnel can verify equipment cleanliness in seconds. Your facility is only as clean as you start, so it's best to start the day right.