Colorimeter
Catalog #: 10752-434
Supplier: Lovibond
Customers Who Bought This Also Bought
Barium chloride dihydrate ≥99.0% ACS, VWR Chemicals BDH®
Supplier: VWR International
$184.95 to $1,807.94
Calcium, Single Element ICP and ICP/MS Certified Reference Standards, Enhanced Packaging, ARISTAR®, VWR Chemicals BDH®
Supplier: VWR International
$194.45
Sulfuric acid 0.1 N in aqueous solution, VWR Chemicals BDH®
Supplier: VWR International
$67.74 to $772.55
Flow cells for MinION devices, Oxford Nanopore Technologies
Supplier: Oxford Nanopore Technologies
$7,900.00
Assurance® Grade Calcium Standards for AA and ICP, SPEX CertiPrep
Supplier: SPEX CERTIPREP LLC
$112.26
Orthophosphoric acid ≥85.0-87.0%, BAKER ANALYZED® ACS, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
$649.00 to $7,523.25