Lovibond MD 100 Colorimeter, Aluminum Range: 0.01 - 0.25 mg/l (Powder Reagents), Reagent included
Catalog #: 10752-430
Supplier: Lovibond
Customers Who Bought This Also Bought
Methanol ≥99.9%, OmniSolv®, gradient grade for HPLC, Supelco®
Supplier: MilliporeSigma
635,20 $ to 2 541,50 $
Ethanol denatured (denatured with 5% isopropanol and 5% methanol), VWR Chemicals BDH®
Supplier: VWR International
147,27 $ to 464,44 $
Single-Use Syringe/BD PrecisionGlide™ Needle Combinations, Sterile, BD Medical
Supplier: BD
46,00 $ to 367,00 $
Water, ARISTAR® ULTRA, Ultrapure for trace metal analysis, VWR Chemicals BDH®
Supplier: BDH
334,23 $ to 2 005,34 $
Iodine standardised solution 0.01 N in aqueous solution
Supplier: Thermo Scientific Chemicals
172,99 $
SystemSURE Plus™ Luminometer, ATP Hygiene Monitoring System, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS CA
1 090,88 $
Acetic acid glacial ≥99.7%, ChromAR® for liquid chromatography, for TLC, Macron Fine Chemicals™
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
195,95 $ to 1 175,58 $