Lovibond MD 100 Colorimeter, HR Chlorine (KI) Range: 5-200 mg/l (Tablet Reagents), Reagent included
Catalog #: 10752-424
Supplier: Lovibond
Customers Who Bought This Also Bought
Acetonitrile ≥99.9% (by GC), OmniSolv®, gradient grade for HPLC, Supelco®
Supplier: MilliporeSigma
684,63 $ to 2 738,51 $
Acetonitrile ≥99.9% (by GC) for HPLC, for spectrophotometry, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
885,46 $ to 3 541,82 $
Blood Agar Plate, 5% Sheep Blood in Tryptic Soy Agar (TSA) Base, 15 × 100 mm plate, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS CA
42,99 $