ContactUsLinkComponent
Vial universal viral transport pk50
BD™ Universal Viral Transport System, BD Medical
Catalog #: CA90006-624
Supplier:  BD
Vial universal viral transport pk50
BD™ Universal Viral Transport System, BD Medical
Catalog #: CA90006-624
Supplier:  BD
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    BD UVT 3 ml Vial
  • Cat. no.:
    CA90006-624
Specifications
About this item
The BD™ Universal Viral Transport System (UVT) is designed to transport viruses, chlamydiae, mycoplasmas, and ureaplasmas at room temperature. It is one of the only systems that may be stored and transported at 2 to 25 °C, all in one formulation.
  • No refrigeration requirements: System does not require refrigerated storage or transport on ice, decreasing the risk of specimen rejection and eliminating messy specimen transport
  • Reduced contamination: System features a unique blend of antimicrobics that target and suppress bacterial and fungal contamination without harming viruses, chlamydiae, mycoplasmas, or ureaplasmas
  • Long-term storage: System helps store samples for long periods by preserving viruses and chlamydiae, when stored frozen with a cryoprotectant
The BD™ Universal Viral Transport System (UVT) is designed to transport viruses, chlamydiae, mycoplasmas, and ureaplasmas at room temperature. It is one of the only systems that may be stored and transported at 2 to 25 °C, all in one formulation.
Customers Who Bought This Also Bought