ASTM Liquid-in-Glass Mercury Thermometers, Partial or Without Immersion, Thermco Products
Catalog #: 10747-822
Supplier: Thermco
Specifications
- Accuracy:0.1 °C
- IP Equivalent:—
- Cat. no.:10747-822
- Divisions:0.1 °C
- ASTM No.:91C
- Application:SOLIDIFICATION POINT
- Range:+20...+50 °C
- Immersion:76 mm
- Supplier No.:ACC091C
- Maximum Length:375 mm
Specifications
About this item
These high precision liquid-in-glass mercury thermometers are hand blown, which bear graduations, numbers and letters etched into the glass and filled with and inert gas above the mercury column
ASTM thermometers are used for a variety of specified tests which quantify and qualify repeatable test results in the fields of oil and petroleum testing.
These thermometers were designed for specific testing and can be calibrated to your specific requirements.
Ordering information: Accessories not included. Certificate of calibration not included. ASTM thermometers with a NIST Traceable Certificate of Calibration from Thermco Products ISO/IEC accredited laboratory are also available; contact your Avantor representative for more information.
Caution: WARNING—Mercury has been designated by EPA and many state agencies as a hazardous material that can cause central nervous system, kidney and liver damage. Mercury, or its vapor, may be hazardous to health and corrosive to materials. Caution should be taken when handling mercury and mercury containing products. See the applicable product Safety Data Sheet (SDS) for details.
Customers Who Bought This Also Bought