Order Entry
ContactUsLinkComponent
ASTM thermometer 2C -5 to 300C, 0, 75, 150, 225, 300C NIST, ISO 17025/ILAC Certified
ISO17025 ASTM Certified Liquid-in-Glass Mercury Thermometers, Thermco Products
Catalog #: 10747-394
Supplier:  Thermco
ASTM thermometer 2C -5 to 300C, 0, 75, 150, 225, 300C NIST, ISO 17025/ILAC Certified
ISO17025 ASTM Certified Liquid-in-Glass Mercury Thermometers, Thermco Products
Catalog #: 10747-394
Supplier:  Thermco

Specifications

  • Accuracy:
    1 °C
  • IP Equivalent:
  • Cat. no.:
    10747-394
  • Divisions:
    1.0 °C
  • ASTM No.:
    2C
  • Application:
    PARTIAL IMMERSION
  • Range:
    −5...+300 °C
  • Immersion:
    76 mm
  • Certification Points:
    0, 75, 150, 225, 300C
  • Supplier No.:
    ACC002CFC
  • Maximum Length:
    395 mm

Specifications

About this item

These high precision liquid-in-glass mercury thermometers are hand blown, which bear graduations, numbers and letters etched into the glass and filled with and inert gas above the mercury column

ASTM thermometers are used for a variety of specified tests which quantify and qualify repeatable test results in the fields of oil & petroleum testing.
WARNING—Mercury has been designated by EPA and many state agencies as a hazardous material that can cause central nervous system, kidney and liver damage. Mercury, or its vapor, may be hazardous to health and corrosive to materials. Caution should be taken when handling mercury and mercury containing products. See the applicable product Safety Data Sheet (SDS) for details.

These thermometers were designed for specific testing and can be calibrated to your specific requirements.

Certifications: A NIST Traceable Certificate of Calibration from Thermco Products ISO/IEC accredited laboratory at the specified certification points.
Ordering information: Accessories not included.
Customers Who Bought This Also Bought