Specifications
- Volume:1 - 20 µl
- Environmentally Preferable:Waste Reducing
- Tip Color:Natural
- Packed:40 Racks, 96 each
- Sterile:+
- Tip Style:Low Binding
- Supplier No.:4136
Specifications
About this item
These low binding barrier tips feature an inert, high-density PE barrier to eliminate aerosol carryover contamination during critical pipetting procedures. The tips will not trap liquids or inhibit PCR in the event of over pipetting.
- Tips are made in a controlled molding environment to ensure accuracy and precision across all tip volumes
- Non pyrogenic
- Human DNA-free, RNase-/DNase-free
- Sterility Assurance Level 10⁻³
This system is designed to help reduce the production and waste of plastic materials. The hinged racks are manufactured with recycled plastic.
Corning DeckWorks barrier tips feature an inert, high-density polyethylene barrier to eliminate aerosol carryover contamination during critical pipetting procedures. DeckWorks tips will not trap liquids or inhibit PCR in the event of over-pipetting.
Corning DeckWorks proprietary low binding technology reduces surface tension on the interior wall of the tip. Leaching and/or subsequent sample degradation associated with silicone-based tips are eliminated. DeckWorks low binding tips are ideal for quantitative assays in which sample binding can significantly alter results. Specific applications include accurate and precise pipetting of expensive reagents, DNA, proteins and/or peptides.
The entire DeckWorks system is comprised of 7 sizes of graduated pipette tips which are longer than standard tips to minimize the contamination issues often associated with traditional pipette tip styles. Sizes include 10, 20, 100, 200, 200, extended length, 300, and 1000 µl tips. The pipette tips are made in a controlled molding environment to ensure accuracy and precision across all tip volumes. All tips are manufactured from polypropylene.