Preserved Rats
Catalog #: 470158-754
Supplier: Avantor
Specifications
- Description:Formalin Preserved Rats
- Size:7 to 9"
- Injection Type:Uninjected
- Cat. no.:470158-754
- Packaging:Vacuum Pack of 10
Specifications
About this item
Low cost and availability make the rat an ideal specimen for mammalian dissection. Rats feature mature reproductive systems and adult musculature. Vacuum packaged.
- Mammalian Anatomy
- General Dissection
- Formalin-Preserved Mink
- In Vacuum Packs
- Plain-Preserved, Single, Double, and Triple-Injected Rats
Our rat specimens are carefully preserved in an extended position, so students to examine their organ systems, and their skeletal, muscular, and reproductive structures.
Teachers can choose from plain preserved, single, double, and triple-injected rats. Injected rats are made without incisions in the neck, ensuring the specimens are intact.
Customers Who Bought This Also Bought