Specifications
- Length:103 mm
- Description:sensION+ Benchtop pH electrode + ATC, 5014T, refillable, double junction with Ag⁺ barrier (high performance)
- Cat. no.:CA89174-198
- pH Range:0 - 14
- Junction:Ceramic diaphragm
- Connector:BNC
- Supplier No.:LZW5014T97002
Specifications
About this item
To meet your analysis needs, combined pH electrodes are available with a variety of lengths, diameters and reference systems.
Our sensION+ high performance pH electrodes have encapsulated reference element, Ag⁺ ion barrier (except 5010T) and are combination type including built in Pt 1000 temperature sensor.
pH glass electrodes from Radiometer Analytical are made with highly shock-resistant glass, making them extremely tough. All pH probes may be used with sensION+ meters.