Order Entry
ContactUsLinkComponent
Funnel powder
VWR® Powder Funnels, Short Stem
Catalog #: 10546-046
Supplier:  VWR International
UL Listed
Funnel powder
VWR® Powder Funnels, Short Stem
Catalog #: 10546-046
Supplier:  VWR International
UL Listed

Specifications

  • Top Diameter:
    80 mm
  • Stem Length:
    30 mm
  • Stem Diameter:
    23 ±2 mm

Specifications

About this item

Funnels comes with short, wide stem, and are useful for adding powder chemical to reaction vessels.

  • Made from ASTM E438 Type 1 Class A borosilicate 33 expansion glass
  • Constructed with a heavy, uniform wall
  • Fire-polished rim and stem to provide long service life

Designed from ASTM Specification E1095 requirements.

Customers Who Bought This Also Bought