VWR® Volumetric Flask with PE Stopper, Class A
Catalog #: 10545-988
Supplier: VWR International
UL Listed
Specifications
- Capacity:25 ml
- Stopper Size:9
- Stopper Type:Polyethylene
- Certification:Batch
Specifications
About this item
These volumetric flasks are batch certified and are calibrated at 20 degrees Celsius To Contain.
- Borosilicate glass fabrication
- Polyethylene stopper construction
- Durable white marking spot
They feature a single graduation ring and polyethylene stopper with marking spot.
Made from ASTM E438 Type 1 Class A Borosilicate 33 expansion glass as per ASTM specification E237 and E288. Durable white color printing which has good life in laboratory environment.
Customers Who Bought This Also Bought