Order Entry
ContactUsLinkComponent
Culture tube
VWR® Culture Tubes without Rim, Reusable
Catalog #: 10545-946
Supplier:  VWR International
UL Listed
Culture tube
VWR® Culture Tubes without Rim, Reusable
Catalog #: 10545-946
Supplier:  VWR International
UL Listed

Specifications

  • Ø:
    18 mm
  • Height:
    150 mm

Specifications

About this item

High-quality test tubes designed and constructed for optimal performance.

  • Fire-polished top
  • No rim
  • Durable white ceramic enamel marking spot

Made from ASTM 438 Type 1 Class A borosilicate 33 expansion glass and as per ASTM specification E982 Type 4.

These tubes come without rim and have fire-polished top. It features a durable white ceramic enamel marking spot.

Allows use at temprature up to 500 °C and have excellent resistance to chemcial attack.

Customers Who Bought This Also Bought