VWR® Round-Bottom Boiling Flasks
Catalog #: 10536-670
Supplier: VWR International
UL Listed
Specifications
- Capacity:500 ml
- Neck Size:24/40
Specifications
About this item
Durable boiling flasks have good temperature and chemical resistance.
- Good temperature resistance
- Chemically resistant
- Uniform wall thickness
Manufactured from ASTM E438 Type 1 Class A borosilicate 33 expansion glass. As per ASTM specification E 1403 Type 1. The flasks feature uniform wall thickness, which makes them ideal for heating.
Customers Who Bought This Also Bought