VWR® Flat-Bottom Boiling Flasks
Catalog #: 10536-648
Supplier: VWR International
UL Listed
Specifications
- Capacity:500 ml
- Neck Size:24/40
Specifications
About this item
The flasks come with uniform wall thickness which makes these flasks ideal for heating.
- Borosilicate glass fabrication
- Good chemical resistance
Durable product has good temperature and chemical resistance.
Manufactured from ASTM E438 Type 1 Class A borosilicate 33 expansion glass. As per ASTM specification E 1403, Type 1 Class 4.
Customers Who Bought This Also Bought