Perform, Nitrile Exam Gloves, Supermax Healthcare Canada
Catalog #: CA10500-782
Supplier: SUPERMAX HEALTHCARE CANADA CA
Specifications
- Ambidextrous/Hand-specific:Ambidextrous
- Glove Color:Green
- Glove size:XS
- Length:24 cm (9 3/8")
- Powdered/Powder-free:Powder-free
- Disposable / reusable:Disposable
- Latex/Latex-free:Latex-free
- Thickness:2.5 mil
- ASTM No.:D6319:00a, EN 455 Part 1
- Finger thickness:0.09 mm
- Palm thickness:0.05 mm
- Cuff thickness:0.05 mm
- Cat. no.:CA10500-782
- Packaging:200 gloves per Dispenser Box
- Supplier No.:92395
Specifications
About this item
Optimal stretchability, just like latex.
- Beaded cuff snugs wrist, eliminates roll-up, minimizes entry of liquids
- Long-lasting comfort, durability and decreased hand fatigue
- Textured fingertips for precision gripping
- Hypoallergenic
- Protect hands from biohazards and a broad range of chemicals
Certifications: ISO 9001: 2000 certified by SGS (UK) and SIRIM (Malaysia); ISO 13485: 2003 certified by SGS (UK); CMDCAS ISO 13485 Certification.
Ordering information: Packaged 200 gloves per dispenser box.
Customers Who Bought This Also Bought