Transform, Nitrile Exam Gloves, Supermax Healthcare Canada
Catalog #: CA10500-560
Supplier: SUPERMAX HEALTHCARE CANADA CA
Specifications
- Ambidextrous/Hand-specific:Ambidextrous
- Glove Color:Blue
- Glove size:XS
- Length:24 cm (9 3/8")
- Powdered/Powder-free:Powder-free
- Disposable / reusable:Disposable
- Latex/Latex-free:Latex-free
- Thickness:3.2 mil
- ASTM No.:D6319:00a, EN 455 Part 1
- Finger thickness:0.10 mm
- Palm thickness:0.07 mm
- Cuff thickness:0.05 mm
- Cat. no.:CA10500-560
- Packaging:100 Gloves per Dispenser Box
- Supplier No.:9889A5
Specifications
About this item
Optimal stretchability, just like latex.
- Beaded cuff snugs wrist, eliminates roll-up, minimizes entry of liquids
- Long-lasting comfort, durability and decreased hand
- Textured fingertips for precision gripping fatigue
- Hypoallergenic
- Protect hands from biohazards and a broad range of chemicals
Certifications: ISO 9001: 2000 certified by SGS (UK) and SIRIM (Malaysia)
ISO 13485: 2003 certified by SGS (UK)
CMDCAS ISO 13485 Certification
ISO 13485: 2003 certified by SGS (UK)
CMDCAS ISO 13485 Certification
Ordering information: Packaged 100 gloves per dispenser box.
Customers Who Bought This Also Bought