Water, Cell Culture Grade low in endotoxins
Catalog #: VWRL0200-0500
Supplier: VWR
CAS Number: 7732-18-5
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Packaging:PETG bottle, clear
- Size:500 ml
- Cat. no.:VWRL0200-0500
- Supplier No.:02-0200-0500
Specifications
Specification Test Results
- Conductivity before packaging:≤ 1.3 uS/cm
- Endotoxin (EQC):≤ 0.25 EU/ml
- Microorganism Growth (EQC):No Growth
- pH (EQC):Passes test
- Total Organic Carbon:< 500 ppb
- Water, Cell Culture Grade low in endotoxins:
Specification Test Results
About this item
Synonyms: Hydrogen oxide
:Water
Boiling Pt:100 °C (1013 hPa)
Melting Pt:0 °C
Density:1 g/cm³ (4 °C)
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Filtered through a 0.2 µm filter.
:Boiling Pt:
:Boiling Pt: 100 °C (1013 hPa)
:Melting Pt:
:Melting Pt: 0 °C
:Flash Pt:
:Flash Pt:
:M.W. 18.02 g/mol
Merck Index:12,10175
EINECS:231-791-2
:0
:100
Storage Temperature:Ambient
:Filter 0,2 um
Customers Who Bought This Also Bought