N,N-Dimethylformamide ≥99.9%, HiPerSolv CHROMANORM® for HPLC, VWR Chemicals BDH®
Catalog #: BDH83634.400
Supplier: VWR International
CAS Number: 68-12-2
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Packaging:Glass bottle, amber
- Size:4 L
- Cat. no.:BDH83634.400
- Supplier No.:BDH83634.400
Specifications
Specification Test Results
- Assay (by GC-FID):Min. 99.9 %
- Water (by KF coulometry):Max. 0.02 %
- Nonvolatile Residue:Max. 0.0005 %
- Acidity:Max. 0.0005 meq/g
- Alkalinity:Max. 0.0002 meq/g
- Transmittance (270 nm):Min. 10 %
- Transmittance (275 nm):Min. 50 %
- Transmittance (290 nm):Min. 80 %
- Transmittance (300 nm):Min. 90 %
- Transmittance (330 nm):Min. 98 %
- Conforms to BDH 15297:Passes test
- N,N-Dimethylformamide ≥99.9%, HiPerSolv CHROMANORM® for HPLC, VWR Chemicals BDH®:
Specification Test Results
About this item
Synonyms: Formic acid dimethylamide,DMFA,Formyldimethylamine,N-Formyldimethylamine,Dimethylformamide,NSC 5356
:Storage Temperature: Ambient
Boiling Pt:153 °C (1013 hPa)
Melting Pt:–61 °C
Density:0.949 g/cm³ (20 °C)
ADR:3,III
:Length:
:Width:
:Height:
:Ø
:Thickness:
:99,9
Flash Pt:57.5 °C (closed cup)
:Boiling Pt:
:Boiling Pt: 153 °C (1013 hPa)
:Melting Pt:
:Melting Pt: –61 °C
:Flash Pt:
:Flash Pt: 57.5 °C (closed cup)
:M.W. 73.09 g/mol
Merck Index:13,03269
EINECS:200-679-5
:153
Storage Temperature:Ambient
UN:2265
REACH:01-2119475605-32
:3
:III
:57.5 °C
:GHS02
:GHS08
:GHS07
Customers Who Bought This Also Bought