Tetrahydrofuran, HiPerSolv CHROMANORM® for HPLC, VWR Chemicals BDH®
Catalog #: BDH28559.100
Supplier: VWR International
CAS Number: 109-99-9
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Packaging:Glass bottle, amber
- Size:1 L
- Cat. no.:BDH28559.100
- Supplier No.:BDH28559.100
Specifications
Specification Test Results
- Assay (by GC-FID) (Corrected for Water):Min. 99.70%
- Water (by KF coulometry):Max. 0.1000%
- Residue after evaporation:Max. 0.0005%
- Acidity:Max. 0.0005 meq/g
- Transmittance at 230 nm:Min. 40%
- Transmittance at 240 nm:Min. 60%
- Transmittance at 250 nm:Min. 70%
- Transmittance at 260 nm:Min. 80%
- Transmittance at 280 nm:Min. 96%
- Transmittance at 300 nm:Min. 98%
- Transmittance (300 nm):Min. 98 %
- Conforms to BDH 15247:Passes test
Specification Test Results
About this item
Synonyms: Butylene oxide,THF
Formula:C₄H₈O
Boiling Pt:66 °C (1013 hPa)
Melting Pt:–108.5 °C
Density:0.888 g/cm³ (20 °C)
Flash Pt:–21.5 °C
MDL Number:MFCD00005356
CAS Number:109-99-9
:Storage Temperature: Ambient
ADR:3,II
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Filtered through a 0.2 µm filter, packaged under nitrogen.
:Boiling Pt:
:Boiling Pt: 66 °C (1013 hPa)
:Melting Pt:
:Melting Pt: –108.5 °C
:Flash Pt:
:Flash Pt: –21.5 °C
:M.W. 72.11 g/mol
Merck Index:12,09351
Storage Temperature:Ambient
EINECS:203-726-8
:66
UN:2056
REACH:05-2114615188-48
:3
:II
:–21.5 °C
:GHS02
:GHS08
:GHS07
:Filter 0,2 um_Nitrogen_filled
Customers Who Bought This Also Bought