Adapter for funnel and with mech. level control , S90 (w) to GL45(m), electrive conductive
Catalog #: 10220-562
Supplier: S.C.A.T.
Adapter for funnel and with mech. level control , S90 (w) to GL45(m), electrive conductive
Catalog #: 10220-562
Supplier: S.C.A.T.
Customers Who Bought This Also Bought
Glycerine ≥99.7%, Laboratory Reagent, VWR Chemicals BDH®
Supplier: VWR International
$881.80 to $2,807.56
Calcium, Single Element ICP and ICP/MS Certified Reference Standards, Enhanced Packaging, ARISTAR®, VWR Chemicals BDH®
Supplier: VWR International
$194.45
Cobalt ≥99.995% (metals basis), wire, Puratronic®, annealed, Ø 1.0 mm (0.04 in)
Supplier: Thermo Scientific Chemicals
$272.99
HyFlex® 11-318 Touch Screen-Capable Gloves, Uncoated, Ansell
Supplier: Ansell Healthcare
$300.30 to $3,498.74