Order Entry
ContactUsLinkComponent
CLOSE-IT 95X95MM GREEN
Control Seal Labels, Bürkle
Catalog #: 10216-986
Supplier:  BURKLE INC
CLOSE-IT 95X95MM GREEN
Control Seal Labels, Bürkle
Catalog #: 10216-986
Supplier:  BURKLE INC

Specifications

  • Color:
    Green
  • Description:
    Printed control seal lables
  • Cat. no.:
    10216-986
  • Dimensions:
    95.3×95 mm (3.75×3.74")
  • Supplier No.:
    5303-0016

Specifications

About this item

Designed to seal the holes in sacks or boxes created when samples are taken, the strong adhesive on the ‘close-it®’ label allows adhesion to surfaces that are slightly moist, frozen or coated with powder. The aluminium backing creates a vapor barrier and seals the container hermetically.

Designed to seal the holes in sacks or boxes created when samples are taken, the strong adhesive on the ‘close-it®’ label allows adhesion to surfaces that are slightly moist, frozen or coated with powder. The aluminium backing creates a vapour barrier and seals the container hermetically. Rolls contain 500 (95×95 mm) or 250 (150×150 mm) stickers.

  • Adheres extremely firmly
  • Can be written on
  • Color coded for easy recognition
Packaging: Rolls contain 500 (95×95 mm) or 250 (150×150 mm) stickers.
Customers Who Bought This Also Bought