Order Entry
ContactUsLinkComponent
NITRATE/NITRITE TEST STRIPS (100)
Quantofix Nitrate/Nitrite Test Strips, CTL Scientific Supply
Catalog #: 10217-528
Supplier:  Macherey-Nagel
CAS Number:  
Quantofix Nitrate/Nitrite Test Strips, CTL Scientific Supply
Catalog #: 10217-528
Supplier:  Macherey-Nagel
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    Nitrate/Nitrite Test Strips
  • Storage:
    Room Temperature
  • Cat. no.:
    10217-528
  • Range:
    NO3 -: 0-10-25-50-100-250-500 mg/L,
    NO2- 0-1-5-10-20-40-80 mg/L NO2-
  • Supplier No.:
    91313

Specifications

About this item

Quantofix Nitrate/Nitrite test strips are for the rapid and reliable determination of Nitrate and Nitrite in solutions

Quantofix Nitrate 100; This test is the highly sensitive version of Quantofix Nitrate/Nitrite. This test allows to detect nitrate down to 5 mg/L. The warning pad for nitrite detects concentrations down to 0.5 mg/L. This test is especially useful for the dairy industry where low levels of nitrate need to be monitored.

Color change is white to red-violet. Quantofix Nitrate 100 has a gradation of scale: 0-5-10-25-50-75-100 mg/L.

The easy dip and read procedure provides a reliable result within 1 minute.

Ordering information: Packaged as a box of 100.
Customers Who Bought This Also Bought