Quantofix Total Iron Test Strips, CTL Scientific Supply
Catalog #: 10217-520
Supplier: Macherey-Nagel
CAS Number:
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Description:Total Iron 1000 Test Strips
- Storage:Room Temperature
- Cat. no.:10217-520
- Range:0-5-20-50-100-250-500-1000 mg/L
- Supplier No.:91330
Specifications
About this item
Quantofix Total Iron strips have necessary reagent already added to its test pad for the detection of Fe2+/3+.
Ordering information: Packaged as a box of 100. Without reagent.
Customers Who Bought This Also Bought