Order Entry
ContactUsLinkComponent
Cell Stimulation Cocktail 500x, Applications: FC, FA
Cell Stimulation Cocktail, Tonbo Biosciences
Catalog #: 10208-548
Supplier:  Tonbo Biosciences
CAS Number:  
Cell Stimulation Cocktail 500x, Applications: FC, FA
Cell Stimulation Cocktail, Tonbo Biosciences
Catalog #: 10208-548
Supplier:  Tonbo Biosciences
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    500X Cell Stimulation Cocktail
  • Size:
    100 µL
  • Cat. no.:
    10208-548
  • Supplier No.:
    TNB-4975-UL100

Specifications

About this item

Cell stimulation cocktail is a ready to use cocktail that can be used for polyclonal cell activation. It is composed of the phorbol ester, PMA (Phorbol 12-Myristate 13-Acetate), a calcium ionophore (Ionomycin), and the protein transport inhibitors Brefeldin A and Monensin. It can be used to elicit cytokine production and retention in a variety of cell types allowing for detection of intracellular protein by flow cytometry.

  • Should be stored at –20 °C
  • To be protected from light and tightly sealed to prevent evaporation

It has been quality-tested by treatment of mouse splenocytes for 6 to 18 hours followed by intracellular staining and flow cytometric analysis of IL-2 or Ifn-gamma. It should be used at 1X final concentration for cell stimulation. It is recommended to add the cocktail directly to the cell suspension at 2 uL/mL to obtain a 1X final concentration.

Application: Flow cytometry and functional assays.

Delivery: Supplied as 500X solution in ethanol.
Customers Who Bought This Also Bought