Order Entry
ContactUsLinkComponent
VWR* Traceable* Dual thermometer 2 bullet probe
VWR® Traceable® Dual Refrigerator/Freezer Thermometers
Catalog #: 10208-396
Supplier:  VWR International
VWR* Traceable* Dual thermometer 2 bullet probe
VWR® Traceable® Dual Refrigerator/Freezer Thermometers
Catalog #: 10208-396
Supplier:  VWR International

Specifications

  • Temperature accuracy:
    +/- 1.0°C
  • Temperature resolution:
    0.1°
  • Probe:
    2 chemically-resistant Bullet™ probes (without bottles)
  • Range:
    50...70 °C (58...158 °F)

Specifications

About this item

Traceable® Dual thermometer displays the temperatures of two separate probes in °C and °F.

  • Easy to operate
  • Resilient against chemicals and impacts
  • Audible and visual alarms

Allows putting one probe in the freezer and another in the refrigerator (or measure two areas of a refrigerator)

High-impact, chemical-resistant ABS plastic case is 3-1/2 x 1-3/4 x 1 inches. Weight is 3-2/3 ounces.

Range is -58 to 158°F and -50 to 70°C with a resolution of 0.1° and an accuracy of ±1°C. Three and a half-foot micro-cable permits refrigerator doors to close on it. Probe is sealed in a miniature bottle (1 x 2-1/2 inches) filled with nontoxic glycol. Solution is GRAS (generally recognized as safe) by the FDA (Food and Drug Administration). Eliminates concerns about incidental contact with food or drinking water. A unit with Bullet™ probes (without bottles) is also available.

Certifications: Traceable to NIST for Accuracy–An individually-numbered Traceable® Certificate is provided which assures accuracy from VWR® ISO 17025 calibration laboratory accredited by A2LA. It indicates traceability to standards provided by NIST (National Institute of Standards and Technology).
Ordering information: Supplied with flip open stand. Velcro® and magnetic strips are provided for mounting of both thermometer and bottle sensor. Supplied with a Traceable® Certificate and AAA battery for continuous, 1-year always-on monitoring.
Customers Who Bought This Also Bought