Order Entry
ContactUsLinkComponent
Filter Tube
ChemScience Filter Tubes with Fritted Disc, Glass Stopcock, SATI International Scientific
Catalog #: CA10201-524
Filter Tube
ChemScience Filter Tubes with Fritted Disc, Glass Stopcock, SATI International Scientific
Catalog #: CA10201-524

Specifications

  • Porosity:
    Coarse
  • Description:
    Filter Tube, Fritted Disc, With 2 mm Glass Stopcock
  • Frit Diameter:
    30 mm
  • ST Joint Size:
    14/20
  • Supplier No.:
    SI-F0351430C

Specifications

About this item

With two 2 mm glass stopcocks having constant pressure retention devices.

Customers Who Bought This Also Bought