TRIS HCl (tris(hydroxymethyl)aminomethane hydrochloride) ≥99.0%, Bioreagent
Catalog #: VWRB85827-1KG
Supplier: VWR
CAS Number: 1185-53-1
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Packaging:Plastic
- Size:1 Kg
Specifications
Specification Test Results
- Appearance:white crystals or crystalline powder
- Assay (By titration):NLT 99.0%
- Absorbance (1M solution) 260nm:NMT 0.05
- Absorbance (1M solution) 280nm:NMT 0.05
- Absorbance (1M solution) 400nm:NMT 0.01
- Identification (by IR):conforms to structure
- Insoluble Matter:NMT 0.005%
- Loss On Drying (at 105ºC):NMT 0.5%
- Melting Point:147⁰C - 153⁰C
- pH (05M sloution in H2O):3.5 to 5.0
- pKa (@ 25ºC):8.0 to 8.4
- Residue On Ignition:NMT 0.1%
- Solubility (1M solution in H2O):clear and complete
- Arsenic (As):NMT 10ppm
- Calcium (Ca):NMT 5 ppm
- Copper (Cu):NMT 5 ppm
- Iron (Fe):NMT 5 ppm
- Lead (Pb):NMT 5 ppm
- Magnesium (Mg):NMT 5 ppm
- Manganese (Mn):NMT 5 ppm
- Zinc (Zn):NMT 5 ppm
- Bioburden:NMT 100 CFU/g
- Cell Culture Toxicity:Passes test
- Endotoxin (10% solution in H2O):NMT 0.1 EU/mg
- DNase:none detected
- RNase:none detected
- Protease:none detected
- TRIS HCl (tris(hydroxymethyl)aminomethane hydrochloride) ≥99.0%, Bioreagent:
Specification Test Results
About this item
Synonyms: Tris(hydroxymethyl)methylamine hydrochloride,2-Amino-2-(hydroxymethyl)propane-1,3-diol hydrochloride,Trizma(R) hydrochloride,Tris(hydroxymethyl)aminomethane hydrochloride,TRIS HCl,Tris(hydroxymethyl)methylammonium chloride,Trometamol hydrochloride
Formula:NH₂C(CH₂OH)₃·HCl
MDL Number:MFCD00012590
:TRIS HCl (Tris(hydroxymethyl) aminomethane hydrochloride)
Melting Pt:150…152 °C
:Length
:Width
:Height
:Diameter
:Thickness
:99,0
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Melting Pt: 150…152 °C
:Flash Pt:
:Flash Pt:
:M.W. 157.6 g/mol
:TRIS HCl (tris(hydroxymethyl)aminomethane hydrochloride)
:150 - 152 °C
:12,09902
Customers Who Bought This Also Bought