ContactUsLinkComponent
Endothelial Cell Basal Medium MV 2: Endothelial Cell Basal Medium MV 2; 500 ml
Endothelial Cell Growth Medium MV, PromoCell
Catalog #: 10175-272
Supplier:  Promocell
CAS Number:  
Endothelial Cell Basal Medium MV 2: Endothelial Cell Basal Medium MV 2; 500 ml
Endothelial Cell Growth Medium MV, PromoCell
Catalog #: 10175-272
Supplier:  Promocell
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    Endothelial Cell Basal Medium MV 2
  • Size:
    500 ml
  • Cat. no.:
    10175-272
  • Supplier No.:
    C-22221
Specifications
About this item
PromoCell Endothelial Cell Growth Medium MV and Endothelial Cell Growth Medium MV2 were developed for the in vitro cultivation of endothelial cells from microvascular vessels, the coronary artery, and the aorta
The media are optimized for primary human cells but can also be used for bovine, porcine, murine, rat and dog endothelial cells from microvascular vessels as well as for various endothelial cell lines.
The media are optimized for primary human cells but can also be used for bovine, porcine, murine, rat and dog endothelial cells from microvascular vessels as well as for various endothelial cell lines.
Recommended for Human Dermal Microvascular Endothelial Cells (HDMEC), Human Cardiac Microvascular Endothelial Cells (HCMEC), Human Coronary Artery Endothelial Cells (HCAEC), Human Aortic Endothelial Cells (HAoEC), Human Pulmonary Microvascular Endothelial Cells (HPMEC), Human Dermal Lymphatic Endothelial Cells (HDLEC), Human Dermal Blood Endothelial Cells (HDBEC).
Endothelial Cell Growth Medium MV2 lacks the Endothelial Cell Growth Supplement (ECGS, bovine hypothalamic extract) but contains the basic Fibroblast Growth Factor, the Insulin-like Growth Factor (Long R3 IGF-1) and the Vascular Endothelial Growth Factor (VEGF). Generally, VEGF leads to higher endothelial cell proliferation in culture. But, because of its multiple effects on the cell metabolism, it may also interfere with certain experimental setups. In these cases, we recommend the use of Endothelial Cell Growth Medium MV.
Ordering information: PromoCell Endothelial Cell Growth Media MV / MV2 are available as Medium (Ready-to-use) or as Medium Kit. The Medium (Ready-to-use) consists of a 500 ml bottle of Basal Medium and one vial of SupplementMix. The Medium Kit consists of a 500 ml bottle of Basal Medium and the SupplementPack (a set of individual vials with pre-measured supplements) allowing the user full control over the media formulation. Adding the SupplementMix or the SupplementPack to the Basal Medium results in the complete Growth Medium. Basal Medium (with or without phenol red) as well as SupplementMix and SupplementPacks can also be purchased separately.
Customers Who Bought This Also Bought