ContactUsLinkComponent
Keratinocyte Growth Medium 2: Keratinocyte Growth Medium 2 (Ready-to-use); 500 ml
Keratinocyte Growth Medium 2, PromoCell
Catalog #: 10175-230
Supplier:  Promocell
CAS Number:  
Keratinocyte Growth Medium 2: Keratinocyte Growth Medium 2 (Ready-to-use); 500 ml
Keratinocyte Growth Medium 2, PromoCell
Catalog #: 10175-230
Supplier:  Promocell
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    Keratinocyte Growth Medium 2 (Ready-to-use)
  • Size:
    500 ml
  • Cat. no.:
    10175-230
  • Supplier No.:
    C-20011
Specifications
About this item
PromoCell Keratinocyte Growth Medium 2 is a serum-free medium developed for the cultivation of epidermal keratinocytes without feeder cells
Recommended for Normal Human Epidermal Keratinocytes (NHEK) juvenile foreskin, single donor, Normal Human Epidermal Keratinocytes (NHEK) juvenile foreskin, pooled, Normal Human Epidermal Keratinocytes (NHEK) adult, single donor, Normal Human Epidermal Keratinocytes (NHEK) adult, pooled.
Recommended for Normal Human Epidermal Keratinocytes (NHEK) juvenile foreskin, single donor, Normal Human Epidermal Keratinocytes (NHEK) juvenile foreskin, pooled, Normal Human Epidermal Keratinocytes (NHEK) adult, single donor, Normal Human Epidermal Keratinocytes (NHEK) adult, pooled.
The medium is optimized for primary human cells but can also be used for horse and porcine keratinocytes, as well as mouse and rat keratinocytes after adapting the CaCl2 concentration. In addition, it is suited for rat corneal keratinocytes.
Ordering information: PromoCell Keratinocyte Growth Medium 2 is available as Medium (Ready-to-use) or as Medium Kit. The Medium (Ready-to-use) consists of a 500 ml bottle of Basal Medium, one vial of CaCl2 stock solution, and one vial of SupplementMix. The Medium Kit consists of a 500 ml bottle of Basal Medium and the SupplementPack (a set of individual vials with pre-measured supplements) allowing the user full control over the media formulation. Adding the Supplement solutions to the Basal Medium results in the complete Growth Medium. Basal Medium (with or without phenol red), SupplementMix as well as SupplementPack can also be purchased separately.
Customers Who Bought This Also Bought