Order Entry
ContactUsLinkComponent
Cryo-SFM: Serum-free medium for cryopreservation; 125 ml
Cryo-SFM Freezing Medium, PromoCell
Catalog #: 10172-414
Supplier:  Promocell
CAS Number:  
Cryo-SFM: Serum-free medium for cryopreservation; 125 ml
Cryo-SFM Freezing Medium, PromoCell
Catalog #: 10172-414
Supplier:  Promocell
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    Cryo-SFM
  • Size:
    125 mL
  • Cat. no.:
    10172-414
  • Supplier No.:
    C-29912

Specifications

About this item

The PromoCell Freezing Medium Cryo-SFM is an animal component-free, chemically defined medium with DMSO, methylcellulose, and other cryoprotective components developed for the cryopreservation of human and animal primary cells

Note Each freeze/thaw cycle leads to a loss in proliferation potential and cell number, particularly when working with fragile primary cells. If freezing the cells, please do so at as low a passage number as possible.

Recommended for Human Primary Cells, Animal Primary Cells, Established Cell Lines, Induced Pluripotent Stem Cells, Embryonic Stem Cells

It ensures high cell viability, excellent growth rates, and good attachment of the cells after thawing. The medium is the first choice for all types of primary cells and cell lines.

Customers Who Bought This Also Bought