To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Description:Fibroblast Growth Medium 2 (Ready-to-use)
- Size:500 ml
- Cat. no.:10172-046
- Supplier No.:C-23020
Specifications
About this item
PromoCell Fibroblast Growth Medium 2 is a low-serum (2% V/V) medium developed for adult human fibroblasts
Recommended for Normal Human Dermal Fibroblasts (NHDF), adult donor, Human Pulmonary Fibroblasts (HPF), Human Aortic Adventitial Fibroblasts (HAoAF), Human Uterine Fibroblasts (HUF), Human Villious Mesenchymal Fibroblasts (HVMF)
Recommended for Normal Human Dermal Fibroblasts (NHDF), adult donor, Human Pulmonary Fibroblasts (HPF), Human Aortic Adventitial Fibroblasts (HAoAF), Human Uterine Fibroblasts (HUF), Human Villious Mesenchymal Fibroblasts (HVMF)
The medium is optimized for primary human cells but can also be used for rat, porcine, and bovine fibroblasts as well as fibroblast cell lines