ContactUsLinkComponent
Fibroblast Growth Medium: Fibroblast Growth Medium (Ready-to-use); 500 ml
Fibroblast Growth Medium for Juvenile Cells, PromoCell
Catalog #: 10172-044
Supplier:  Promocell
CAS Number:  
Fibroblast Growth Medium: Fibroblast Growth Medium (Ready-to-use); 500 ml
Fibroblast Growth Medium for Juvenile Cells, PromoCell
Catalog #: 10172-044
Supplier:  Promocell
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    Fibroblast Growth Medium (Ready-to-use)
  • Size:
    500 ml
  • Cat. no.:
    10172-044
  • Supplier No.:
    C-23010
Specifications
About this item
The PromoCell Fibroblast Growth Medium is a serum-free medium optimized for the cultivation of neonatal and juvenile fibroblasts
Recommended for Normal Human Dermal Fibroblasts (NHDF), juvenile foreskin
The medium is optimized for primary human cells but can also be used for rat, porcine, and bovine fibroblasts as well as fibroblast cell lines
Ordering information: The Medium (Ready-to-use) consists of a 500 ml bottle of Basal Medium and one vial of SupplementMix. The Medium Kit consists of a 500 ml bottle of Basal Medium and the SupplementPack (a set of individual vials with pre-measured supplements) allowing the user full control over the media formulation. Adding the SupplementMix or the SupplementPack to the Basal Medium results in the complete Growth Medium. Basal Medium (with or without phenol red) as well as the SupplementMix and SupplementPack can also be purchased separately.
Customers Who Bought This Also Bought